Learn More
Biomatik Corporation Canine C-Reactive Protein, Crp Elisa Kit
Supplier:  Biomatik Corporation EKC31845
Encompass_Preferred
Canine C-Reactive Protein, Crp Elisa Kit for detecting C-Reactive Protein, Crp in serum, plasma, cell culture supernates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.312-20ug/ml. Sensitivity: 0.078ug/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: T2KEN6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7231
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.