Learn More
Biomatik Corporation Goat Immunoglobulin A(IgA) ELISA Kit
Supplier: Biomatik Corporation EKC32201
Sample Type: serum, plasma. Species Reactivity: Capra hircus (Goat). Detection Range: 29 ng/mL-7500 ng/mL. Sensitivity: 29 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.