Learn More
Biomatik Corporation Human Claudin-3 (Cldn3) Elisa Kit
Supplier:  Biomatik Corporation EKC33146
Encompass_Preferred
Human Claudin-3 (Cldn3) Elisa Kit for detecting Claudin 3 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O15551; CLDN3. Target Synonyms: CLDN3; C7orf1; CPETR2; Claudin-3; Clostridium perfringens enterotoxin receptor 2; CPE-R 2; CPE-receptor 2; Rat ventral prostate.1 protein homolog; hRVP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7364
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.