Learn More
Biomatik Corporation Human Lipoprotein, a (Lpa) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05666
The Human Lipoprotein, a (Lpa) ELISA Kit allows for quantitative determination of Lipoprotein, a in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.48 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08519. Cross Activity: No significant cross-reactivity or interference between Lipoprotein, a and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3148