Learn More
Biomatik Corporation Human TJP1 (Tight junction protein ZO-1) ELISA Kit
Supplier:  Biomatik Corporation EKF59374
Encompass_Preferred
Human TJP1 (Tight junction protein ZO-1) ELISA Kit for detecting TJP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q07157. Target Synonyms: Tight junction protein 1; Tight junction protein ZO 1; TJP1; ZO 1; ZO1; ZO-1; Zona occludens protein 1; Zonula occludens protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8262
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.