Learn More
Biomatik Corporation Human pro-IL1β (pro interleukin 1 beta) ELISA Kit
Supplier:  Biomatik Corporation EKF60427
Encompass_Preferred
Human pro-IL1β (pro interleukin 1 beta) ELISA Kit for detecting pro-IL1β in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01584. Target Synonyms: pro-IL1β. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8347
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.