Learn More
Biomatik Corporation Mouse Gm6484 (Hepatocellular carcinoma-associated protein TD26 homolog) ELISA Kit
Supplier:  Biomatik Corporation EKF60774
Encompass_Preferred
Mouse Gm6484 (Hepatocellular carcinoma-associated protein TD26 homolog) ELISA Kit for detecting Gm6484 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8R1L8. Target Synonyms: Gm6484; Hepatocellular carcinoma-associated protein TD26 homolog; ANGPTL8; Betatrophin; C19orf80; Gm6484; Lipasin; PRO1185; PVPA599; RIFL; TD26; C19orf80; chromosome 19 open reading frame 80; hepatocellular carcinoma-associated gene TD26; hepatocellular carcinoma-associated protein TD26; PRO1185; PVPA599; TD26; UNQ599; Hepatocellular carcinoma-associated protein TD26 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8441
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.