Learn More
Biomatik Corporation Human Mannose Associated Serine Protease 2 (MASP2) ELISA Kit
Supplier:  Biomatik Corporation EKL58913
Encompass_Preferred
Human Mannose Associated Serine Protease 2 (MASP2) ELISA Kit for detecting MASP2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: O00187. Gene ID: 10747. Target Synonyms: Mannose-binding protein-associated serine protease 2; mannan-Binding Lectin Serine Peptidase 2; MBL-associated serine protease 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8733
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.