Learn More
Biomatik Corporation Rat Tpsab1 (Tryptase,Mast cell protease 7) ELISA Kit
Supplier:  Biomatik Corporation EKF58756
Encompass_Preferred
Rat Tpsab1 (Tryptase,Mast cell protease 7) ELISA Kit for detecting Tpsab1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P27435. Target Synonyms: Tpsab1; Tryptase; skin; Mast cell protease 7; rMCP-7; Tryptase alpha; beta-1;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8181