Learn More
Biomatik Corporation Canine BNP (Brain Natriuretic Peptide) ELISA Kit
Supplier:  Biomatik Corporation EKF60739
Encompass_Preferred
Canine BNP (Brain Natriuretic Peptide) ELISA Kit for detecting BNP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P16859. Target Synonyms: BNP; Brain Natriuretic Peptide; NPPB; GC-B; B-Type Natriuretic Peptide; Natriuretic Peptide Precursor B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8418
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.