Learn More
Biomatik Corporation Human Plasminogen Activator Inhibitor 1 (PAI1) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU06658
The Human Plasminogen Activator Inhibitor 1 (PAI1) ELISA Kit allows for quantitative determination of Plasminogen Activator Inhibitor 1 in Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.5-800 pg/mL. Sensitivity: 5.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05121. Cross Activity: No significant cross-reactivity or interference between Plasminogen Activator Inhibitor 1 and analogues was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.