Learn More
Biomatik Corporation Mouse Mitochondrial Open Reading Frame Of The 12S rRNA-c (MOTS-c) ELISA Kit
Supplier:  Biomatik Corporation EKU11280
Encompass_Preferred
Mouse Mitochondrial Open Reading Frame Of The 12S rRNA-c (MOTS-c) ELISA Kit for detecting MOTS-c in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 246.9-20,000pg/ml. Sensitivity: 98.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6882
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.