Learn More
Biomatik Corporation Human Protein C (PROC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06892
Encompass_Preferred
The Human Protein C (PROC) ELISA Kit allows for quantitative determination of Protein C in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 29.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04070. Cross Activity: No significant cross-reactivity or interference between Protein C and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3633