Learn More
Biomatik Corporation Mouse Proteinase 3 (PR3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06959
Encompass_Preferred
Mouse Proteinase 3 (PR3) ELISA Kit for detecting PR3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61096. Gene ID: 19152. Target Synonyms: MBN; PRTN3; ACPA; AGP7; C-ANCA; MBT; P29; PRK; Myeloblastin; Serine Proteinase,Neutrophil,Wegener Granulomatosis Autoantigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3653
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.