Learn More
Biomatik Corporation Fish lysozyme (renal amyloidosis) (LZM) ELISA kit
Supplier:  Biomatik Corporation EKC32163
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Fish. Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 1.56 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7274
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.