Learn More
Biomatik Corporation Human Regulated On Activation In Normal T-Cell Expressed And Secreted (RANTES) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU07053
Human Regulated On Activation In Normal T-Cell Expressed And Secreted (RANTES) ELISA Kit for detecting RANTES in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13501. Gene ID: 6352. Target Synonyms: CCL5; SCYA5; SISd; EoCP; Chemokine C-C-Motif Ligand 5; Eosinophil Chemotactic Cytokine; T-cell Specific Protein p288; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.