Learn More
Biomatik Corporation Human Podocin Elisa Kit
Supplier:  Biomatik Corporation EKC40706
Encompass_Preferred
Human Podocin Elisa Kit for detecting Nephrosis 2, Idiopathic, Steroid-Resistant (Podocin) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NP85; NPHS2. Target Synonyms: nephrosis 2; Nephrosis 2; idiopathic; steroid resistant (podocin); nephrosis 2; idiopathic; steroid resistant; NPHS2; NPHS2 gene; PDCN; PODO_HUMAN; Podocin; SRN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7770
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.