Learn More
Biomatik Corporation Rat MT (Melatonin) ELISA Kit
Supplier:  Biomatik Corporation EKE62000
Encompass_Preferred
Rat MT (Melatonin) ELISA Kit for detecting MT in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h, 30min. Target Synonyms: N-Acetyl-5-Methoxytryptamine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8533
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.