Learn More
Biomatik Corporation Human NACHT, LRR and PYD Domains-Containing Protein 3(NLRP3) ELISA Kit
Supplier:  Biomatik Corporation EKL59410
Encompass_Preferred
Human NACHT, LRR and PYD Domains-Containing Protein 3(NLRP3) ELISA Kit for detecting NLRP3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.068ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: Q96P20. Gene ID: 114548. Target Synonyms: PYRIN-containing APAF1-like protein 1; Cryopyrin; Cold autoinflammatory syndrome 1 protein; Angiotensin/vasopressin receptor AII/AVP-like; Caterpiller protein 1.1(CLR1.1). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8750
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.