Learn More
Biomatik Corporation Human Procollagen III N-Terminal Propeptide (PIIINP) ELISA Kit
Supplier:  Biomatik Corporation EKL59896
Encompass_Preferred
Human Procollagen III N-Terminal Propeptide (PIIINP) ELISA Kit for detecting PIIINP in serum, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 25.3pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Target Synonyms: P3NP; N-Propeptide Of Type III Procollagen; Procollagen III Amino Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8762
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.