Learn More
Biomatik Corporation Human Regenerating Islet Derived Protein 1 Alpha (REG1a) ELISA Kit
Supplier:  Biomatik Corporation EKU07038
Encompass_Preferred
Human Regenerating Islet Derived Protein 1 Alpha (REG1a) ELISA Kit for detecting REG1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05451. Gene ID: 5967. Target Synonyms: REG; PSP; ICRF; PSPS1; PTP; Islet cells regeneration factor; Islet of Langerhans regenerating protein; Lithostathine 1 Alpha; Pancreatic Stone Protein; Pancreatic Thread Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6353
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.