Learn More
Biomatik Corporation Triglyceride (TG) ELISA Kit
Supplier:  Biomatik Corporation EKU08730
Encompass_Preferred
Triglyceride (TG) ELISA Kit for detecting TG in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Universal. Detection Range: 3.9-1,000ug/ml. Sensitivity: 1.8ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: TAG; Triacylglycerol; Triacylglyceride. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6535
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.