Learn More
Biomatik Corporation Cyclic Guanosine Monophosphate (cGMP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08690
Encompass_Preferred
The Cyclic Guanosine Monophosphate (cGMP) ELISA Kit allows for quantitative determination of Cyclic Guanosine Monophosphate in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 123.5-10000 pg/mL. Sensitivity: 45.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Cyclic Guanosine Monophosphate and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4262