Learn More
Biomatik Corporation Rat Gasdermin D (GSDMD) ELISA Kit
Supplier:  Biomatik Corporation EKU09199
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.062ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: A0A096MJ11. Target Synonyms: GSDMDC1; DF5L; Gasdermin Domain Containing 1. Restriction: For Research Use Only.
Catalog No. 50-228-6590