Learn More
Biomatik Corporation Indole 3 Acetic Acid (IAA) CLIA Kit
Supplier:  Biomatik Corporation EKU08638
Encompass_Preferred
Sample Type: Tissue or cell culture supernates. Species Reactivity: General species. Detection Range: 1.56-400ng/mL. Sensitivity: 0.59ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Competitive. Assay Time: 2 hours. UniProt ID: Not Available. Target Synonyms: IA-A; Indolylacetic Acid; Indoleacetic Acid; Heteroauxin. Restriction: For Research Use Only.
Catalog No. 50-228-6520
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.