Learn More
Biomatik Corporation Dog Superoxide Dismutases (SOD) ELISA Kit
Supplier:  Biomatik Corporation EKU11445
Encompass_Preferred
Dog Superoxide Dismutases (SOD) ELISA Kit for detecting SOD in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6927
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.