Learn More
Biomatik Corporation Human Lactate Dehydrogenase A (Ldha) Elisa Kit
Supplier:  Biomatik Corporation EKC34361
Encompass_Preferred
Human Lactate Dehydrogenase A (Ldha) Elisa Kit for detecting Lactate Dehydrogenase A in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000mU/ml. Sensitivity: 7.81mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00338; LDHA. Target Synonyms: Cell proliferation-inducing gene 19 protein; GSD11; L lactate dehydrogenase B chain; L-lactate dehydrogenase A chain; Lactate dehydrogenase A; Lactate dehydrogenase B; Lactate dehydrogenase c variant 1; Lactate dehydrogenase c variant 3; Lactate dehydrogenase c variant 4; Lactate dehydrogenase C4; Lactate dehydrogenase H chain; Lactate dehydrogenase M; LDH A; LDH B; LDH H; LDH heart subunit; LDH M; LDH muscle subunit; LDH-A; LDH-M; LDH1; ldha; LDHA_HUMAN; LDHBD; LDHM; MS1111; PIG19; Proliferation inducing gene 19; Proliferation-inducing gene 19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7464
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.