Learn More
Biomatik Corporation Human Programmed Death Ligand-1 (Pd-L1/Cd274) Elisa Kit
Supplier:  Biomatik Corporation EKC35128
Encompass_Preferred
Human Programmed Death Ligand-1 (Pd-L1/Cd274) Elisa Kit for detecting Cd274 Molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NZQ7; CD274. Target Synonyms: B7 H; B7 H1; B7 homolog 1; B7-H1; B7H; B7H1; CD 274; CD274; CD274 antigen; CD274 molecule; MGC142294; MGC142296; OTTHUMP00000021029; PD L1; PD-L1; PD1L1_HUMAN; PDCD1 ligand 1; PDCD1L1; PDCD1LG1; PDL 1; PDL1; Programmed cell death 1 ligand 1; Programmed death ligand 1; RGD1566211. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7526
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.