Learn More
Biomatik Corporation Rabbit Bone Alkaline Phosphatase (BALP) ELISA Kit
Supplier:  Biomatik Corporation EKC38402
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 0.156 mIU/mL-10 mIU/mL. Sensitivity: 0.039 mIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: G1SLU6. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7641