Learn More
Biomatik Corporation Human Mucin-5Ac (Muc5Ac) Elisa Kit
Supplier:  Biomatik Corporation EKC34663
Encompass_Preferred
Human Mucin-5Ac (Muc5Ac) Elisa Kit for detecting Mucin-5Ac in serum, plasma, cell culture supernates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P98088; MUC5AC. Target Synonyms: MUC5AC; MUC5; Mucin-5AC; MUC-5AC; Gastric mucin; Major airway glycoprotein; Mucin-5 subtype AC; tracheobronchial; Tracheobronchial mucin; TBM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7485
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.