Learn More
Biomatik Corporation Mouse circulating immune complex(CIC)ELISA Kit
Supplier:  Biomatik Corporation EKC36559
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7584
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.