Learn More
Biomatik Corporation Human Angiogenin, Ang Elisa Kit
Supplier:  Biomatik Corporation EKC40028
Encompass_Preferred
Human Angiogenin, Ang Elisa Kit for detecting Angiogenin, Ribonuclease, Rnase A Family, 5 in serum, plasma, cell culture supernates, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P03950; ANG. Target Synonyms: ALS9; AMYOTROPHIC LATERAL SCLEROSIS; ANG 1; ANG; ANG I; ANG1; ANGI; ANGI_HUMAN; Angiogenin; Angiogenin ribonuclease RNase A family; 5; Epididymis luminal protein 168; HEL168; MGC22466; MGC71966; Ribonuclease 5; Ribonuclease RNase A Family 5; RNase 5; RNASE4; RNASE5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7690
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.