Learn More
Biomatik Corporation Indole 3 Acetic Acid (IAA) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11626
Encompass_Preferred
Indole 3 Acetic Acid (IAA) Mini sample ELISA Kit for detecting IAA in tissue or cell culture supernates. Species Reactivity: Universal. Detection Range: 320-200,000pg/ml. Sensitivity: 112pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: IA-A; Indolylacetic Acid; Indoleacetic Acid; Heteroauxin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6975