Learn More
Biomatik Corporation Human Hyaluronic Acid, Ha Elisa Kit
Supplier:  Biomatik Corporation EKC34084
Encompass_Preferred
Human Hyaluronic Acid, Ha Elisa Kit for detecting Hyaluronic Acid, Ha in serum, plasma, cerebrospinal fluid (csf), tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7436
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.