Learn More
Biomatik Corporation Bovine Immunoglobulin A, IgA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31661
Encompass_Preferred
The Bovine Immunoglobulin A, IgA ELISA Kit allows for quantitative determination in serum, plasma, cell culture supernates, tissue homogenates. Detection Range: 0.39 μg/mL-25 μg/mL. Sensitivity: 0.39 μg/mL. Format: 96-Well Plate. Assay Type: ELISA. Restriction: For Research Use Only.
Catalog No. 50-149-7495
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.