Learn More
Biomatik Corporation Pig oxytocin,OT ELISA Kit
Supplier:  Biomatik Corporation EKC38301
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 2.5 pg/mL-100 pg/mL. Sensitivity: 2.5 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: MGC126890, MGC126892, OT, OT-NPI, neurophysin I|oxytocin, prepro- (neurophysin I)|oxytocin-neurophysin I|oxytocin-neurophysin I, preproprotein. Restriction: For Research Use Only.
Catalog No. 50-228-7636
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.