Learn More
Biomatik Corporation Human Granulocyte-Macrophage Colony Stimulating Factor, Gm-Csf Elisa Kit
Supplier:  Biomatik Corporation EKC40051
Encompass_Preferred
Human Granulocyte-Macrophage Colony Stimulating Factor, Gm-Csf Elisa Kit for detecting Colony Stimulating Factor 2 (Granulocyte-Macrophage) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04141; CSF2. Target Synonyms: Colony stimulating factor 2 (granulocyte-macrophage); Colony Stimulating Factor 2; Colony stimulating factor; Colony-stimulating factor; CSF 2; CSF; CSF2; CSF2_HUMAN; GM-CSF; GMCSF; Granulocyte Macrophage Colony Stimulating Factor; Granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGI1GM; Molgramostin; Pluripoietin-a; Sargramostim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7697
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.