Learn More
Biomatik Corporation Human Mesothelin (Msln) Elisa Kit
Supplier:  Biomatik Corporation EKC40992
Encompass_Preferred
Human Mesothelin (Msln) Elisa Kit for detecting Mesothelin(Msln) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13421; MSLN. Target Synonyms: CAK 1; CAK1; CAK1 antigen; cleaved form; Megakaryocyte potentiating factor; Mesothelin; Mesothelin isoform 1 precursor; MPF; Msln; MSLN_HUMAN; Pre pro megakaryocyte potentiating factor; Pre-pro-megakaryocyte-potentiating factor; SMR; SMRP; Soluble MPF mesothelin related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7808
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.