Learn More
Biomatik Corporation Human WNT5A (Protein Wnt-5a) ELISA Kit
Supplier:  Biomatik Corporation EKF57312
Encompass_Preferred
Human WNT5A (Protein Wnt-5a) ELISA Kit for detecting WNT5A in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P41221. Target Synonyms: WNT5A; hWNT5A; Protein Wnt-5a; wingless-type MMTV integRation site family; member 5A; WNT-5A protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7919
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.