Learn More
Biomatik Corporation Human Mortalin (75 kDa glucose-regulated protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58929
Encompass_Preferred
Human Mortalin (75 kDa glucose-regulated protein) ELISA Kit for detecting Mortalin in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P38646. Target Synonyms: CSA; GRP 75; GRP75; Heat shock 70 kDa protein 9; HSPA9; HSPA9B; Mortalin; MOT; mot 2; MOT2; MTHSP75; PBP74; Peptide binding protein 74. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8212
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.