Learn More
Biomatik Corporation Total MMP-1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51986
Encompass_Preferred
Total MMP-1 (Human) Sandwich ELISA Kit for detecting Total MMP-1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 157-10000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P03956. Gene ID: 4312. Target Synonyms: Interstitial collagenase; Fibroblast collagenase; Matrix metalloproteinase-1; CLG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9161
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.