Learn More
Biomatik Corporation TNF-alpha (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51917
Encompass_Preferred
TNF-alpha (Murine) Sandwich ELISA Kit for detecting TNF-alpha in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06804. Gene ID: 21926. Target Synonyms: Tumor necrosis factor; Cachectin; TNF-alpha; Tumor necrosis factor ligand superfamily member 2; TNFSF2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9135
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.