Learn More
Biomatik Corporation AF10 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50783
Encompass_Preferred
AF10 Colorimetric Cell-Based ELISA for detecting AF10 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P55197. Gene ID: 8028. Target Synonyms: Protein AF-10; MLLT10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1007
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.