Learn More
Biomatik Corporation MAEA Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50892
Encompass_Preferred
MAEA Colorimetric Cell-Based ELISA for detecting MAEA in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q7L5Y9. Gene ID: 10296. Target Synonyms: Erythroblast macrophage protein; Cell proliferation-inducing gene 5 protein; Lung cancer-related protein 10; HLC-10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1111