Learn More
Biomatik Corporation TSLP (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52006
Encompass_Preferred
TSLP (Murine) Sandwich ELISA Kit for detecting TSLP in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9JIE6. Gene ID: 53603. Target Synonyms: Thymic stromal lymphopoietin; Thymic stroma-derived lymphopoietin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1934
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.