Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 28 (ADAM28) ELISA Kit
Supplier:  Biomatik Corporation EKU02049
Encompass_Preferred
Human A Disintegrin And Metalloprotease 28 (ADAM28) ELISA Kit for detecting ADAM28 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UKQ2. Gene ID: 10863. Target Synonyms: ADAM23; MDCL; eMDCII; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L; Epididymal metalloproteinase-like, disintegrin-like, and cysteine-rich protein II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2047
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.