Learn More
Biomatik Corporation Mouse Toll Like Receptor 2 (TLR2) ELISA Kit
Supplier:  Biomatik Corporation EKU07748
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9QUN7. Target Synonyms: CD282; TIL4; Toll/interleukin-1 receptor-like protein 4. Restriction: For Research Use Only.
Catalog No. 50-229-2948
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.