Learn More
Biochain Institute Inc ELISA Kit for IgM Antibody to Hepatitis A Virus, 1 kit/PK
SDP
Supplier:  Biochain Institute Inc KO31008096
Encompass_Preferred
BioChain offers an extensive list of ELISA kits for research use of detection of infectious diseases, such as HBV, HCV, and HIV. The kits are approved and used in clinical diagnosis in other countries, such as China. The assay is a conventional immunological assay where purified antibody is coated on the solid phase of multi-wells. Serum sample, antigen and horseradish peroxidase conjugated antibodies are added to coated wells. The test is special, sensitive, reproducible and easy to operate.
Catalog No. 50-180-834
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.