Learn More
Description
The ELISA Plate-coating Buffer quantifies Buffers, Chelators and Reagents.
Specifications
Specifications
| For Use With (Application) | ELISA |
| Product Line | R&D Systems |
| Quantity | 250 mL |
| Solution Type | Buffer |
| Grade | Not Available |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
