Learn More
ProteinSimple Simple Reader™ ELISA Plate Reader
Supplier:  ProteinSimple IA051123
Encompass
Empowering Precision with Ease and Simplicity
Empower every lab member with their own Simple Reader – a groundbreaking microplate reader featuring 96 individual detection units, ensuring each person has their own compact, reliable device for decentralized workflows. Needing just a laptop, its plug-and-play functionality seamlessly transforms any location into your lab space, making fieldwork a breeze.
Compact Design: Take this personal plate reader on the go to any lab, anywhere.
4 LED Light Sources: 450 nm, 490 nm, 540 nm, 595 nm
96 Detection Units: Robust, reliable results every time.
Fast Read Time: Speed up data analysis by reading your plate in under 5 seconds.
Catalog No. 50-000-14336
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.